GC Analysis of Small Molecules in Jet Fuel on Supel-Q™ PLOT
GC Analysis of Small Molecules in Jet Fuel on Supel-Q™ PLOT
Conditions
column Supel-Q PLOT
oven 35 °C (3 min), 16 C/min to 250 °C
carrier gas helium, 3.0 mL/min
sample Jet fuel
detector TCD
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Petroleum
Legal Information Supel-Q is a trademark of Sigma-Aldrich Co. LLC