Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Solvents on a Packed Column (3% SP-1500 on Carbopack™ B)
GC Analysis of Solvents on a Packed Column (3% SP-1500 on Carbopack™ B)
Conditions
column 10 ft (3.048m) x 1/8 in.(9.52 mm) stainless steel
packing Carbopack(TM) B/3% SP(TM)-1500
column temp. A & C - 70 C to 225 C at 4 C/min B - 100 C to 225 C at 8 C/min
carrier gas nitrogen, 20 ml/min
inj. temp. 0.3 muL3% each component by volume (approx. 12 mug each)
detector FID
Description
Categories Analytical Chromatography, GC Applications, Solvents
Featured Industry Chemicals and Industrial Polymers
Legal Information Carbopack is a trademark of Sigma-Aldrich Co. LLC, SP is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 2-Nitropropane
  • Acetone
  • Butyl Cellosolve
  • Cellosolve acetate
  • Cyclohexanone
  • Ethanol
  • Ethyl acetate
  • Ethyl benzene
  • Ethyl Cellosolve
  • Ethyl formate
  • Isoamyl acetate
  • Isobutyl acetate
  • Isobutyl alcohol
  • Isopropyl alcohol
  • Mesityl oxide
  • Methanol
  • Methyl acetate
  • Methyl Cellosolve
  • Methyl ethyl ketone
  • Methyl formate
  • Methylene chloride
  • m-Xylene
  • n-Butyl acetate
  • n-Butyl alcohol
  • n-Propyl acetate
  • n-Propyl alcohol
  • o-Xylene
  • p-Xylene
  • sec-Butyl acetate
  • sec-Butyl alcohol
  • Tetrahydrofuran (THF)
  • Toluene