GC Analysis of Solvents on a Packed Column (1%SP-2410 on Carbopack™ B)
GC Analysis of Solvents on a Packed Column (1%SP-2410 on Carbopack™ B)
Conditions
column 10 ft(3.048m) x 1/8 in. (9.82 mm), stainless steel
packing Carbopack™ B/1% SP™-1510 (11809)
column temp. A & C - 70C to 225C at 4C/min B - 100C to 225C at 8C/min
carrier gas nitrogen, 20 mL/min
inj. temp. 0.3 µL3% each component by volume (approx. 12 mug each)
detector FID
Description
Categories Analytical Chromatography, GC Applications, Solvents
Featured Industry Chemicals and Industrial Polymers
Legal Information Carbopack is a trademark of Sigma-Aldrich Co. LLC, SP is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes