GC Analysis of Spearmint Essential Oil on SLB®-IL59, 80 °C Initial Oven Temp.
GC Analysis of Spearmint Essential Oil on SLB®-IL59, 80 °C Initial Oven Temp.
Conditions
column SLB-IL59, 30 m x 0.25 mm I.D., 0.20 µm (28891-U)
oven 80 °C, 3 °C/min to 280
carrier gas helium, 40 cm/sec
sample spearmint essential oil diluted 1:10 in hexane
injection 1 µL100:1 split
inj. temp. 250 °C
detector FID, 305 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes