Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Sterols (Silylated Derivatives) in Olive Oil (Free Sterol Fraction) on SLB®-5ms after SPE using Discovery DSC-Si
GC Analysis of Sterols (Silylated Derivatives) in Olive Oil (Free Sterol Fraction) on SLB®-5ms after SPE using Discovery DSC-Si
Conditions
sample preparation SPE (Solid Phase Extraction)
sample/matrix olive oil spiked with a mixed sterol standard at 5 µg/g
SPE tube/cartridge Discovery DSC-Si, 1 g/6 mL (52656-U)
condition 2 x 5 mL hexane
sample addition 1 mL
elution esters fraction: 2.5 mL of hexane:ethyl acetate (90:10); free sterols fraction; 5 mL of hexane:ethyl acetate (90:10) and 3 mL x 5 of ethanol:diethyl ether:hexane (50:25:25)
eluate post-treatment Evaporate to dryness and perform silylation as described in the Analysis Note.
column SLB-5ms, 20 m x 0.20 mm I.D., 0.20 µm (28564-U)
oven 125 C (1 min), 10 C/min to 325 C (10 min)
carrier gas helium, 0.6 ml/min, constant
sample Olive oil sample; fractionated, transesterified, and silylated
injection 1 µL, splitless
liner 4 mm I.D. single taper
inj. temp. 250 C
detector MSD, rn/z 500 - 600
MSD interface 325 C
Description
Analysis Note Sterol content is one in a battery of tests used to analyze the composition and determine the grade and authenticity of olive oil. Test results are compared against standards such as those established by the International Olive Oil Council (IOC) and US Dept. of Agriculture. In vegetable oils, sterols exist in the free form, or esterified to fatty acids. In this application, sterols were analyzed in olive oil separately as free and esterified. This was accomplished by taking advantage of the polarity difference between these two groups to fractionate them using silica gel SPE. In place of saponification, the fractions were transesterified to liberate the esterified sterols. The fractions were then silylated, and analyzed by GC-MS. The silylated sterols all contained a significant molecular ion in their mass spectum, which made it possible to perform peak identification free of matrix interference. The sterols isolated in the free fraction are shown here., Sample pre-treatment
Weigh 0.25 gm oil into test tube. For spiked sample add 12.5 μL of 65-100 μg/mL mixed sterol std. Add 1 ml of hexane:ethyl acetate (90:10). After SPE fractionation, evaporate solvent from each fraction, add 1 ml of 30% methanolic sodium methoxide:MeOH:MTBE solution (13:27:60), mix and let sit at room temp for 30 min. Add 1 ml of water and 2 mL of n-heptane. Withdraw aqueous phase (bottom) and replace w/ 1 mL of 1% citric acid solution. Withdraw organic phase and dry over anhydrous sodium sulfate. Silylation: Evaporate organic phase to dryness. Add 125 μL of Sylon BFT and 125 μL of pyridine. Heat at 70°C for 20 min. The sample is then ready for GC-MS analysis.
Categories Analytical Chromatography, SPE Applications, GC Applications, Sterols
Featured Industry Agriculture
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC, application for SPE
Analyte
  • Brassicasterol
  • Campesterol
  • Cholesterol
  • Stigmasterol
  • β-Sitosterol