GC Analysis of Sugars (as Alditol Acetates) on Equity®-1701
GC Analysis of Sugars (as Alditol Acetates) on Equity®-1701
Conditions
column Equity-1701, 30 m x 0.25 mm I.D., 0.25 µm (28372-U)
oven 220 °C
carrier gas helium, 20 cm/sec @ 220 °C
sample Alditol acetate mixes @ 500 ppm in methylene chloride (47880-U & 47881)
injection 1 µL25:1 split
inj. temp. 220 °C
detector FID, 260 °C
Description
Categories Analytical Chromatography, GC Applications, Alditol Acetates
Featured Industry Food and Beverages
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes