GC Analysis of Sulfur Gases on Supel-Q™ PLOT
GC Analysis of Sulfur Gases on Supel-Q™ PLOT
Conditions
column Supel-Q PLOT, 30 m x 0.53 mm I.D. (25462)
oven 50 °C (1 min), 10 °C/min to 250 °C (10 min)
carrier gas helium, 3 mL/min
sample 7-Component sulfur compound mix, analytes at concentrations indicated
injection 25 µL
inj. temp. 230 °C
detector FPD, 260 °C
Description
Categories Analytical Chromatography, GC Applications, Volatiles, Sulfur Compounds, Gases
Featured Industry Chemicals and Industrial Polymers
Legal Information Supel-Q is a trademark of Sigma-Aldrich Co. LLC