GC Analysis of TCEP Test Mix on SLB®-IL100, 80 °C Oven Temp.
GC Analysis of TCEP Test Mix on SLB®-IL100, 80 °C Oven Temp.
Conditions
column SLB-IL100, 30 m x 0.25 mm I.D., 0.20 µm (28884-U)
oven 80 C
carrier gas helium, 30 cm/sec
sample TCEP Test Mix (N08682), each analyte at various concentrations in isooctane
injection FID, 250 C
injection 1.0 muL100:1 split
liner 4 mm I.D., split, cup design
inj. temp. 250 C
Description
Categories Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons
Featured Industry Petroleum
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,2,4,5-Tetramethylbenzene
  • 1,2,4-Trimethylbenzene
  • Cyclohexanone
  • Ethylbenzene
  • Isooctane
  • Isopropylbenzene (cumene)
  • n-Tridecane
  • p-Xylene
  • Toluene