Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of TCEP Test Mix on SLB®-IL100, 110 °C Oven Temp.
GC Analysis of TCEP Test Mix on SLB®-IL100, 110 °C Oven Temp.
Conditions
column SLB-IL100, 30 m x 0.25 mm I.D., 0.20 µm (28884-U)
oven 110 C
carrier gas helium, 30 cm/sec
sample (TCEP Test Mix (N08682), each analyte at various concentrations in isooctane)
injection 1.0 muL100:1 split
liner 4 mm I.D., split, cup design
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons
Featured Industry Petroleum
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,2,4,5-Tetramethylbenzene
  • 1,2,4-Trimethylbenzene
  • Cyclohexanone
  • Ethylbenzene
  • Isooctane
  • Isopropylbenzene (cumene)
  • n-Tridecane
  • p-Xylene
  • Toluene