GC Analysis of Terpenes in Hops (Cascade Variety) on Equity®-1 after Headspace SPME using 50/30 μm DVB/CAR/PDMS Fiber
GC Analysis of Terpenes in Hops (Cascade Variety) on Equity®-1 after Headspace SPME using 50/30 μm DVB/CAR/PDMS Fiber
Conditions
sample preparation 30 min equilibration at 40 °C prior to extraction
sample preparation fiber postbake after extraction, 3 min at 270 °C
sample/matrix ground Cascade variety hop pellets
desorption process 3 min, 270 °C
extraction 20 min, headspace, 40 °C
SPME fiber Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), 50/30 μm (57298-U)
column Equity-1, 60 m x 0.25 mm I.D., 0.25 µm (28047-U)
oven 60 °C (2 min), 5 °C/min to 275 °C (5 min)
carrier gas helium, 1 mL/min constant flow
liner 0.75 mm ID, SPME
inj. temp. 270 °C
detector MSD
MSD interface 300 °C
scan range full scan, m/z 50-500
Description
Analysis Note Terpenes are contained in the essential oils derived from plants, and often impart characteristic aromas to the plant and/or oil. In this application, headspace-SPME combined with GC/MS was used to analyze some of the terpenes present in the Cascade variety of hops. Chromatographic separation was performed on an Equity®-1 capillary GC column, and identification was done using retention indices and spectral library match. Analysis showed a characteristic terpene profile, with high levels of ß-myrcene, caryophyllene, and humulene present.
Categories Analytical Chromatography, GC Applications, Terpenes, Hydrocarbons
Featured Industry Food and Beverages
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes add 0.5 gm ground hops to a 10 mL headspace vial
suitability application for SPME, application for GC