GC Analysis of Terpinen-4-ol and Methyl Mandelate Enantiomers on α-DEX 120, β-DEX 120, and γ-DEX 120
GC Analysis of Terpinen-4-ol and Methyl Mandelate Enantiomers on α-DEX 120, β-DEX 120, and γ-DEX 120
Conditions
column 30 m x 0.25 mm I.D., 0.25 µm (alpha-DEX 24310, beta-DEX 24304, gamma-DEX 24307)
oven 130 °C
carrier gas helium, 35 cm/sec
sample 1 mg/mL in methylene chloride
injection 1 µL100:1 split
inj. temp. 250 °C
detector FID, 300 °C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Terpenes, Alcohols
Featured Industry Chemicals and Industrial Polymers
Flavors and Fragrances
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • (-)-Terpinen-4-ol
  • (+)-Terpinen-4-ol
  • R(-)-Methyl mandelate
  • S(+)-Methyl mandelate