Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms
GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 45 C (3 min), 20 C/min to 360 C (10 min)
carrier gas helium, 1.3 ml/min. constant
sample TPH Mix 3, each analyte at 1000 µg/mL in carbon disulfide (861394-U)
injection 1.0 µL, 100:1 split
liner 2 mm I.D. straight
inj. temp. 275 C
detector FID, 365 C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Environmental
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Carbon disulfide
  • Decane
  • Dodecane
  • Dotriacontane
  • Eicosane
  • Heptane
  • Hexadecane
  • Hexane
  • Hexatriacontane
  • Nonane
  • Octacosane
  • Octadecane
  • Octane
  • Tetracontane
  • Tetracosane
  • Tetradecane
  • Tetratetracontane
  • Undecane