GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Conditions
column SLB-5ms, 20 m x 0.18 mm I.D., 0.18 µm (28564-U)
oven 70 C (0.5 min), 20 C/min to 360 C (1 min)
carrier gas hydrogen, 2.5 ml/min constant
sample TPH Mix 3, each analyte at 1000 µg/mL in carbon disulfide (861394-U)
injection 0.50 µL, 200:1 split
liner 2 mm I.D. straight
inj. temp. 275 C
detector FID, 365 C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Environmental
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Carbon disulfide
  • Decane
  • Dodecane
  • Dotriacontane
  • Eicosane
  • Heptane
  • Hexadecane
  • Hexane
  • Hexatriacontane
  • Nonane
  • Octacosane
  • Octadecane
  • Octane
  • Tetracontane
  • Tetracosane
  • Tetradecane
  • Tetratetracontane
  • Undecane