GC Analysis of Verbenone Enantiomers on Astec® CHIRALDEX® G-TA
GC Analysis of Verbenone Enantiomers on Astec® CHIRALDEX® G-TA
Conditions
column Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 µm (73033AST)
oven 120 °C
carrier gas helium, 30 psi
sample 2.5 mg/mLpeak 1: R(+)-verbenonepeak 2: S(-)-verbenone
injection 1 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Can reverse elution order by using CHIRALDEX B-TA.
Categories Analytical Chromatography, GC Applications, Enantiomers, Terpenes
Featured Industry Chemicals and Industrial Polymers
Flavors and Fragrances
Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes