GC Analysis of Volatiles on SPB®-624, Direct Injection
GC Analysis of Volatiles on SPB®-624, Direct Injection
Conditions
column SPB-624, 60 m x 0.25 mm I.D., 1.4 µm (24256)
oven 35 C (4 min), 4 °C/min to 200 °C
carrier gas helium, 33 cm/sec @ 40 °C
sample 5 ng on column of each component
injection 70:1 split
inj. temp. 250 °C
scan range m/z 45-300
Description
Categories Analytical Chromatography, GC Applications, Volatiles
Featured Industry Environmental
Food and Beverages
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,1,1,2-Tetrachloroethane
  • 1,1,1-Trichloroethane
  • 1,1,2,2-Tetrachloroethane
  • 1,1,2-Trichloroethane
  • 1,1-Dichloroethane
  • 1,1-Dichloroethene
  • 1,1-Dichloropropene
  • 1,2,3-Trichlorobenzene
  • 1,2,3-Trichloropropane
  • 1,2,3-Trimethylbenzene
  • 1,2,4-Trichlorobenzene
  • 1,2,4-Trimethylbenzene
  • 1,2-Dibromo-3-chloropropane
  • 1,2-Dibromoethane
  • 1,2-Dichlorobenzene
  • 1,2-Dichloroethane
  • 1,2-Dichloropropane
  • 1,3-Dichlorobenzene
  • 1,3-Dichloropropane
  • 1,4-Dichlorobenzene
  • 2,2-Dichloropropane
  • 2-Chlorotoluene
  • 4-Chlorotoluene
  • Benzene
  • Bromobenzene
  • Bromochloromethane
  • Bromodichloromethane
  • Bromoform
  • Bromomethane
  • Carbon tetrachloride
  • Chlorobenzene
  • Chloroethane
  • Chloroform
  • Chloromethane
  • cis-1,2-Dichloroethene
  • cis-1,3-Dichloropropene
  • Dibromochloromethane
  • Dibromomethane
  • Dichlorodifluoromethane
  • Ethylbenzene
  • Hexachlorobutadiene
  • Isopropylbenzene (cumene)
  • Methylene chloride
  • m-Xylene
  • Naphthalene
  • n-Butylbenzene
  • n-Propylbenzene
  • o-Xylene
  • p-Cymene
  • p-Isopropyltoluene
  • sec-Butylbenzene
  • Styrene
  • tert-Butylbenzene
  • Tetrachloroethene
  • Toluene
  • trans-1,2-Dichloroethene
  • trans-1,3-Dichloropropene
  • Trichloroethene
  • Trichlorofluoromethane
  • Vinyl chloride