GC Analysis of Volatiles in Irish Whiskey on VOCOL® after SPME using 50/30 μm DVB/Carboxen®/PDMS Fiber
GC Analysis of Volatiles in Irish Whiskey on VOCOL® after SPME using 50/30 μm DVB/Carboxen®/PDMS Fiber
Conditions
sample preparation Solid Phase Extraction
sample/matrix 4 mL Irish whiskey (80 proof) in a 10 mL vial (no salt added)
desorption process 270 °C for 3 min
extraction 50/30 µm divinylbenzene/Carboxen®/polydimethylsiloxane on StableFlex™ (57298-U)
SPME fiber 50/30 µm divinylbenzene/Carboxen®/polydimethylsiloxane on StableFlex™ (57298-U)
column VOCOL®, 30 m x 0.25 mm I.D., 1.50 µm (24205-U)
oven 40 °C (2.5 min), 6 °C/min to 55 °C, 45 °C/min to 235 °C (5min)
carrier gas helium, 1.0 mL/min
injection splitless (closed 0.75 min)
liner 0.75 mm I.D., direct (SPME) type, straight design
inj. temp. 270 °C
detector FID, 300 °C
Description
Analysis Note Beverages are liquid, so sample preparation is often simple, and can be accomplished with direct injection, headspace, solid phase microextraction (SPME), solid phase extraction (SPE), or liquid-liquid extraction (LLE). SPME coupled with GC is an effective technique for monitoiring the volatiles present in the headspace over spirits.
Categories Analytical Chromatography, SPME Applications, GC Applications, Food and Beverage, Alcohols, Volatiles
Featured Industry Food and Beverages
Legal Information StableFlex is a trademark of Sigma-Aldrich Co. LLC, Carboxen is a registered trademark of Sigma-Aldrich Co. LLC, VOCOL is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for SPME, application for GC