GC Analysis of Volatiles in Milk Thistle Honey on SUPELCOWAX® 10 after Dynamic Headspace (DHS) using 20/35 Tenax® TA Tube
GC Analysis of Volatiles in Milk Thistle Honey on SUPELCOWAX® 10 after Dynamic Headspace (DHS) using 20/35 Tenax® TA Tube
Conditions
sample/matrix 25 g milk thistle honey in a 200 mL Erlenmeyer flask at 45 C, purged with nitrogen at 60 mL/min for 30 min as a stripping gas to collect onto a Tenax® TA tube
dry purge back-flushed with nitrogen, 60 mL/min at room temperature for 5 min
thermal desorption tube 20/35 Tenax® TA, 90 mg
desorption process helium, 10 mL/min at 280 °C for 10 min to cryofocus onto a glass-lined tube at -120 °C, which was subsequently heated to 240 °C for transfer to the GC column
column SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.25 µm (24079)
oven 35 C (8 min), 6 C/min to 60 °C, 4 °C/min to 160 °C, 20 °C/min to 200 °C (1 min)
carrier gas helium, 1 mL/min
transfer line/valve temp. 250 °C, transfer line directly coupled to the GC column using a press-fit connector
MSD interface 230 °C
scan range 35-300 m/z
Description
Categories Analytical Chromatography, Air Monitoring Applications, GC Applications, Volatiles
Featured Industry Agriculture
Food and Beverages
Legal Information SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC, Tenax is a registered trademark of Buchem B.V.
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for air monitoring, application for GC
Analyte
  • 3-Phenylfuran
  • Benzaldehyde
  • Decanal
  • Furfural
  • Hotrienol
  • Nonanal
  • Nonanoic acid
  • Nonanol
  • Phenylacetaldehyde
  • Phenylethyl alcohol
  • α-Linalool