Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Volatiles in Multifloral Honey on SUPELCOWAX® 10 after SPME using a 50/30 μm DVB/Carboxen/PDMS Fiber
GC Analysis of Volatiles in Multifloral Honey on SUPELCOWAX® 10 after SPME using a 50/30 μm DVB/Carboxen/PDMS Fiber
Conditions
sample/matrix 5 g multifloral honey + 5 mL bidistilled water in a 20 mL vial, allowed to equilibrate at 50 C for 15 min with magnetic stirring
desorption process 250 C for 2 min
extraction headspace, 50 C for 40 min with magnetic stirring
SPME fiber 2 cm 50/30 µm divinylbenzene/carboxen on polydimethylsiloxane (57348-U)
column SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.25 µm (24079)
oven 35 C (8 min), 6 C/min to 60 C, 4 C/min to 160 C, 20 C/min to 200 C (1 min)
carrier gas helium, 1 ml/min
sample (Honey)
liner 0.75 mm I.D., SPME
inj. temp. 250 C
MSD interface 230 C
scan range 35-300 m/z
Description
Analysis Note In this application, headspace SPME (HS-SPME) using a Carboxen/PDMS/DVB fiber coupled with gas chromatography-mass spectrometry (GC/MS) was used for the characterization of the volatile fraction of honey samples. Using SPME it was possible to obtain and distinguish the chemical fingerprints of different types of honey. The relatively polar SUPELCOWAX 10 GC column provides excellent separation of these polar compounds.
Categories Analytical Chromatography, SPME Applications, GC Applications, Agriculture, Foods, Flavors & Fragrances, Volatiles
Featured Industry Agriculture
Food and Beverages
Legal Information SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for SPME, application for GC
Analyte
  • 3-Phenylfuran
  • Benzaldehyde
  • Decanal
  • Furfural
  • Hotrienol
  • Nonanal
  • Nonanoic acid
  • Nonanol
  • Phenylacetaldehyde
  • Phenylethyl alcohol
  • α-Linalool