GC Analysis of Volatiles in Purple Nitrile Gloves on SPB®-HAP after Collection/Desorption using Carbotrap® 300 Tube
GC Analysis of Volatiles in Purple Nitrile Gloves on SPB®-HAP after Collection/Desorption using Carbotrap® 300 Tube
Conditions
sample/matrix 0.5 g purple nitrile glove
dry purge 15 min
thermal desorption tube Carbotrap® 300 Glass-Fritted Thermal Desorption Tube, 1/4 in O.D. x 3.5 in L (30141-U)
desorption process 350 °C with 10 min hold
column SPB®-HAP, 60 m x 0.32 mm I.D., 4 µm (25020-U)
oven 40 °C (3 min), 12 °C/min to 200 °C (20 min)
carrier gas helium
transfer line/valve temp. 180 °C
detector MSD
MSD interface 210 °C
Description
Categories Analytical Chromatography, Air Monitoring Applications, GC Applications, Volatiles
Featured Industry Industrial Hygiene
General description Qualitative volatile headspace outgassing from purple nitrile gloves using the ATIS system with analysis by thermal desorption.
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC, Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for air monitoring, application for GC
Analyte
  • 4-Cyanocyclohexene
  • 5-Dodecene
  • 6-Methyl-5-hepten-2-one
  • Carbon disulfide
  • Diethyl cyanamide
  • Methacrylic acid
  • N,N-Diethylformamide
  • Naphthalene