Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Volatiles in White Wine on VOCOL® after SPME using a 75 μm Carboxen/PDMS Fiber
GC Analysis of Volatiles in White Wine on VOCOL® after SPME using a 75 μm Carboxen/PDMS Fiber
Conditions
sample/matrix white wine + 25% NaCl
desorption process 3 min at 290 C
extraction 10 min, 40 C (headspace)
SPME fiber Carboxen/PDMS, 75 µm (57318)
column VOCOL, 30 m x 0.25 mm I.D., 1.5 µm (24205-U)
oven 40 C (2 min) to 220 C at 10 C/min
carrier gas helium, 35 cm/sec
sample white wine
injection splitless/split, closed 1 min
liner 0.75 mmliner
inj. temp. 290 C
detector GC-MS, quadrupole, m/z = 31-240
Description
Analysis Note In this application, SPME was used for the analysis of volatiles in white wine using a headspace method. The presence of aroma compounds in white wine is one of the most important attributes of its final quality. A Carboxen/PDMS fiber was inserted into the headspace of the vial for 10 minutes at 40 °C and analyzed by GC/MS. The thick film of the VOCOL GC column helps focus the volatile analytes.
Categories Analytical Chromatography, SPME Applications, GC Applications, Food & Beverage Analysis, Foods, Flavors & Fragrances, Volatiles, Beverage Testing, Beer, Wine, and Spirits, Wine
Featured Industry Food and Beverages
Legal Information VOCOL is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for SPME, application for GC
Analyte
  • 2,3-Butanediol
  • 2-Methyl-1-butanol
  • Acetic acid
  • Ethanol
  • Ethyl acetate
  • Ethyl butyrate
  • Ethyl hexanoate
  • Ethyl octanoate
  • Hexanol
  • Hexyl acetate
  • Isoamyl alcohol
  • Isobutanol
  • Isopentanol
  • Methyl formate
  • Octanoic acid
  • Sulfur dioxide