GC Analysis of Water (0.01-0.5%Calibration Curve) on Watercol™ 1460
GC Analysis of Water (0.01-0.5%Calibration Curve) on Watercol™ 1460
Conditions
column Watercol 1460, 30 m x 0.25 mm I.D., 0.20 µm (29695-U)
oven 60 °C (12 min)
carrier gas helium, 26 cm/sec
sample 5 standards, water at 0.01, 0.05, 0.1, 0.3, and 0.5% (v/v), each in ethanol
injection 0.5 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector TCD, 200 °C
Description
Analysis Note The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water can be integrated and quantified. This data shows the linearity obtained from a 5-point calibration curve (0.01-0.5%) for water on Watercol 1460.
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Flavors and Fragrances
Food and Beverages
Petroleum
Pharmaceutical (small molecule)
Legal Information Watercol is a trademark of Sigma-Aldrich Co. LLC