GC Analysis of Water (100%Standard) on Watercol™ 1460
GC Analysis of Water (100%Standard) on Watercol™ 1460
Conditions
column Watercol 1460, 30 m x 0.25 mm I.D., 0.20 µm (29695-U)
oven 60 C (12 min)
carrier gas helium, 26 cm/sec
sample water
injection 0.1 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 C
detector TCD, 200 C
Description
Analysis Note The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water can be integrated and quantified. This application shows a calibration standard (100% water) on Watercol 1460.
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Flavors and Fragrances
Food and Beverages
Petroleum
Pharmaceutical (small molecule)
Legal Information Watercol is a trademark of Sigma-Aldrich Co. LLC