GC Analysis of Wieland-Miescher Ketone Enantiomers on Astec® CHIRALDEX® G-BP
GC Analysis of Wieland-Miescher Ketone Enantiomers on Astec® CHIRALDEX® G-BP
Conditions
column Astec CHIRALDEX G-BP, 10 m x 0.25 mm I.D., 0.12 µm (75031AST)
oven 155 °C
carrier gas helium, 12 psi
sample peak 1: R(-)-Wieland-Miescher Ketonepeak 2: S(+)-Wieland-Miescher Ketone
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Elution order reversal on CHIRALDEX G-TA and CHIRALDEX G-DP
Categories Analytical Chromatography, GC Applications, Ketones, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes