Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GCxGC Analysis of C4-C24 FAMEs on SLB®-5ms x SLB-IL60, Non-Polar to Polar Strategy
GCxGC Analysis of C4-C24 FAMEs on SLB®-5ms x SLB-IL60, Non-Polar to Polar Strategy
Conditions
column (1D) SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
column (2D) SLB-IL60, 1 m x 0.10 mm I.D., 0.08 µm
oven (1D) 40 C (2 min), 4 C/min to 270 C (20 min)
oven (2D) 55 C (2 min), 4 C/min to 280 C (20 min)
oven (modulator) temp. offset 30 C, modulation period 3 sec
carrier gas helium, 1.0 ml/min
sample C4-C24 FAME mix
injection 1 muL20:1 split
inj. temp. 250 C
detector TOFMS, mass range 35-450 m/z, acquisition rate 200 spectra/sec, ion source temp. 250 C
Description
Analysis Note LECO Pegasus 4D GCxGC/TOFMS
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC