GPA Method 2261: GC Analysis of Natural Gas on a Packed Column (GPA 3-Column Set)
GPA Method 2261: GC Analysis of Natural Gas on a Packed Column (GPA 3-Column Set)
Conditions
column GPA 2261 3-column set (13993-U)
oven 100 C
carrier gas helium, 30 ml/min
injection 0.5 mLsample loop
Description
Analysis Note Packed columns continue to be used in multi-column, multi-valve configurations for the determination of BTU content in natural gas and natural gas liquids as described in Gas Processors Association (GPA) Methods 2261 and 2177, plus American Society for Testing and Materials (ASTM) Test Method 2597. This chromatogram is of GPA Method 2261, and uses three packed columns in a multi-valve configuration. Supelco is the world leader in packed GC column technology, with an extensive library of phases and supports.
Categories Analytical Chromatography, GC Applications, Volatiles, Gases, Hydrocarbons
Featured Industry Petroleum
Other Notes Complementary products:
Syringes