OSHA Method 62: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SPB®-20 after Collection/Desorption using ORBO™-49P
OSHA Method 62: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SPB®-20 after Collection/Desorption using ORBO™-49P
Conditions
adsorbent tube ORBO-49P (20350)
column SPB-20, 60 m x 0.53 mm I.D., 1.0 µm film (available on request)
oven 200 °C (2 min) to 250 °C at 4 °C/min
carrier gas helium, 8 mL/min
sample calibration standard: 12 µg/mL each pesticide, 78 µg/mL tributyl phosphate (int. std.) in hexane (blank: 270 mg Supelpak 20, glass fiber filter desorbed in toluene + 78 µg/mL tributyl phosphate.)
injection 1 µLdirect injection, 220 °C (packed column injector)
detector FPD, 300 °C
Description
Categories Analytical Chromatography, GC Applications, Air Monitoring Applications, Pesticides
Featured Industry Industrial Hygiene
General description This is an application, not a product for sale.
See "Related Products" to find associated products for sale.
Or, for more information, email: Supelco Technical Service
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC, ORBO is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for air monitoring, application for GC
Analyte
  • Chlorpyrifos
  • Diazinon
  • Dichlorvos
  • Malathion
  • Parathion
  • Tributyl phosphate