Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
SFC Analysis of 1-(1-Naphthyl)ethylamine Enantiomers on Astec® Cellulose DMP
SFC Analysis of 1-(1-Naphthyl)ethylamine Enantiomers on Astec® Cellulose DMP
Conditions
column Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 µm particles (51098AST)
column temp. 40 C
mobile phase 20% methanol (0.1% MSA, methanesulfonic acid) in CO2
flow rate 3 ml/min
pressure 1740 psi (120 bar)
sample 2 mg/mL in methanol
injection 5 µl
detector UV, 230 nm
Description
Categories Analytical Chromatography, SFC Applications
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for SFC