SFC Analysis of Diastereomers on Astec® Cellulose DMP, Methanol Additive
SFC Analysis of Diastereomers on Astec® Cellulose DMP, Methanol Additive
Conditions
column Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 µm particles (51098AST)
column temp. 35 °C
mobile phase [A] CO2; [B] methanol; (80:20, A:B)
flow rate 3 mL/min
pressure 1450 psi (100 bar)
injection 5 µL
detector UV, 220 nm
Description
Analysis Note This application shows the ability of Astec Cellulose DMP to separate a mixture of diastereomers. The analytes are proprietary pharmaceutical-type compounds.
Categories Analytical Chromatography, SFC Applications
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes SFC data provided by Dr. Christina Kraml, Lotus Separations, LLC, Princeton, NJ. Instrument was a Berger Analytical SFC system equipped with a dual pump (FCM-1200), autosampler (ALS 3100), column oven (TCM-200) and diode-array detector (DAD-4100); from Mettler Toledo.
suitability application for SFC