Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
SFC Analysis of Flavanone Enantiomers on Astec® Cellulose DMP, Methanol Additive
SFC Analysis of Flavanone Enantiomers on Astec® Cellulose DMP, Methanol Additive
Conditions
column Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 µm particles (51098AST)
column temp. 35 C
mobile phase [A] CO2; [B] isopropanol; (80:20, A:B)
flow rate 3 ml/min
pressure 1450 psi (100 bar)
sample flavanone, 2 mg/mL
injection 5 µl
detector UV, 220 nm
Description
Analysis Note SFC data provided by Dr. Christina Kraml, Lotus Separations, LLC, Princeton, NJ.
Instrument: Berger Analytical SFC system equipped with a dual pump (FCM-1200), autosampler (ALS 3100), column oven (TCM-200) and diode-array detector (DAD-4100); from Mettler Toledo.
Categories Analytical Chromatography, SFC Applications
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for SFC