US EPA Method 0030: GC Analysis of Volatiles on SPB®-624 after Collection/Desorption using VOST 100 Tube
US EPA Method 0030: GC Analysis of Volatiles on SPB®-624 after Collection/Desorption using VOST 100 Tube
Conditions
adsorbent tube VOST 100 (20074-U)
purge trap VOCARB 4000 (20308)
column SPB-624, 75 m x 0.53 mm I.D., 3.0 µm film (25432)
oven 35 °C (5 min) to 200 °C at 10 °C/min, hold 2 min
carrier gas helium, 8 mL/min
sample calibration standard: volatiles in methanol; blank: Tenax TA tube with surrogate thermally desorbed into purge & trap system
detector FID, 300 °C
Description
Categories Analytical Chromatography, GC Applications, Air Monitoring Applications, Volatiles
Featured Industry Environmental
Industrial Hygiene
General description This is an application, not a product for sale.
See "Related Products" to find associated products for sale.
Or, for more information, email: Supelco Technical Service
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for air monitoring, application for GC
Analyte
  • 1,1,1-Trichloroethane
  • 1,1,1-Trichloropropane
  • Benzene
  • Chlorobenzene-d5
  • Methanol
  • Methylene chloride
  • Toluene