US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, ELCD Signal
US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using
Conditions
sample/matrix 5 mL water (5 ppb each analyte)
purge trap VOCARB 3000 (21131-U)
purge 37 mL/min for 11 min at 25 C
desorption process 220 °C for 0.5 min
column VOCOL, 105 m x 0.53 mm I.D., 3.0 µm (25358)
oven 35 C (10 min), 4 °C/min to 200 °C
carrier gas helium, 10 mL/min (plus 20 mL/min as make-up gas)
detector ELCD/PID in series
Description
Analysis Note Sample Preparation: Purge and trap using a VOCARB 4000 trap (8.5cm Carbopack™ C/10cm Carbopack B/6cm Carboxen 1000/1cm Carboxen 1001).
GC Column: VOCOL wide bore capillary column, 105m x 0.53mm ID fused silica, 3.0μm phase film.
Detector: Photoionization and electrolytic/conductivity, in series.
Categories Analytical Chromatography, GC Applications, Volatiles
Featured Industry Environmental
Food and Beverages
Legal Information VOCOL is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC, application for SP
Analyte
  • 1,1,1,2-Tetrachloroethane
  • 1,1,1-Trichloroethane
  • 1,1,2,2-Tetrachloroethane
  • 1,1,2-Trichloroethane
  • 1,1-Dichloroethane
  • 1,1-Dichloroethene
  • 1,1-Dichloroethylene
  • 1,1-Dichloropropane
  • 1,1-Dichloropropene
  • 1,2,3-Trichlorobenzene
  • 1,2,3-Trichloropropane
  • 1,2,4-Trichlorobenzene
  • 1,2,4-Trimethylbenzene
  • 1,2-Dibromo-3-chloropropane
  • 1,2-Dibromoethane
  • 1,2-Dichlorobenzene
  • 1,2-Dichloroethane
  • 1,2-Dichloropropane
  • 1,3,5-Trimethylbenzene
  • 1,3-Dichlorobenzene
  • 1,3-Dichloropropane
  • 1,4-Dichlorobenzene
  • 2,2-Dichloropropane
  • 2-Chlorotoluene
  • 4-Chlorotoluene
  • Benzene
  • Bromobenzene
  • Bromochloromethane
  • Bromodichloromethane
  • Bromoform
  • Bromomethane
  • Carbon tetrachloride
  • Chlorobenzene
  • Chloroethane
  • Chloroform
  • Chloromethane
  • cis-1,2-Dichloroethylene
  • cis-1,3-Dichloropropene
  • Dibromomethane
  • Dichlorodifluoromethane
  • Ethylbenzene
  • Hexachlorobutadiene
  • Isopropylbenzene (cumene)
  • Methylene chloride
  • m-Xylene
  • Naphthalene
  • o-Xylene
  • p-Cymene
  • Propylbenzene
  • p-Xylene
  • sec-Butylbenzene
  • Styrene
  • tert-Butylbenzene
  • Tetrachloroethylene
  • Toluene
  • trans-1,2-Dichloroethylene
  • trans-1,3-Dichloropropene
  • Trichloroethylene
  • Trichlorofluoromethane
  • Vinyl chloride