US EPA Method 524.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap
US EPA Method 524.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using
Conditions
purge trap VOCARB 3000 (21066-U)
purge 40 mL/min for 11 min
dry purge 3 min
desorption process 250 C for 4 min
bake 280 °C for 10 min
column VOCOL, 105 m x 0.53 mm I.D., 3.0 mum (25358)
oven 35 C (10 min), 4 C/min to 200 C (10 min)
carrier gas helium, 10 mL/min
sample 10 ppb each component in 5 mL water
scan range m/z = 35-260 at 0.6 scan/sec
Description
Categories Analytical Chromatography, GC Applications, Volatiles
Featured Industry Environmental
Food and Beverages
Legal Information VOCOL is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,1,1,2-Tetrachloroethane
  • 1,1,1-Trichloroethane
  • 1,1,2,2-Tetrachloroethane
  • 1,1,2-Trichloroethane
  • 1,1-Dichloroethane
  • 1,1-Dichloroethylene
  • 1,1-Dichloropropene
  • 1,2,3-Trichlorobenzene
  • 1,2,3-Trichloropropane
  • 1,2,4-Trichlorobenzene
  • 1,2,4-Trimethylbenzene
  • 1,2-Dibromo-3-chloropropane
  • 1,2-Dibromoethane
  • 1,2-Dichlorobenzene
  • 1,2-Dichlorobenzene-d4
  • 1,2-Dichloroethane
  • 1,2-Dichloropropane
  • 1,3,5-Trimethylbenzene
  • 1,3-Dichlorobenzene
  • 1,3-Dichloropropane
  • 1,4-Dichlorobenzene
  • 2,2-Dichloropropane
  • 2-Chlorotoluene
  • 4-Chlorotoluene
  • Benzene
  • Bromobenzene
  • Bromochloromethane
  • Bromodichloromethane
  • Bromoform
  • Bromomethane
  • Carbon tetrachloride
  • Chlorobenzene
  • Chlorodibromomethane
  • Chloroethane
  • Chloroform
  • Chloromethane
  • cis-1,2-Dichloroethylene
  • cis-1,3-Dichloropropene
  • Dibromomethane
  • Dichlorodifluoromethane
  • Ethylbenzene
  • Fluorobenzene
  • Hexachlorobutadiene
  • Isopropylbenzene (cumene)
  • Methylene chloride
  • m-Xylene
  • Naphthalene
  • n-Butylbenzene
  • n-Propylbenzene
  • o-Xylene
  • p-isopropyltoluene
  • p-Xylene
  • sec-Butylbenzene
  • Styrene
  • tert-Butylbenzene
  • Tetrachloroethylene
  • Toluene
  • trans-1,2-Dichloroethylene
  • trans-1,3-Dichloropropene
  • Trichloroethylene
  • Trichlorofluoromethane
  • Vinyl chloride