US EPA Method 608.1/608.2: GC Analysis of Organochlorine Pesticides on Equity®-1701
US EPA Method 608.1/608.2: GC Analysis of Organochlorine Pesticides on Equity®-1701
Conditions
column Equity-1701, 30 m x 0.25 mm I.D., 0.25 µm (28372-U)
oven 75 °C (2 min), 15 °C/min to 280 °C (5 min)
carrier gas helium, 1.2 mL/min. constant
sample mixture of 608.1 and 608.2 calibration solutions (40351-U and 40352-U) diluted to 100 ppb in n-hexane
injection 1 µL, splitless (splitter open after 0.75 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
detector ECD, 300 °C
Description
Categories Analytical Chromatography, GC Applications, Pesticides
Featured Industry Environmental
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes