US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SPB®-608
US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SPB®-608
Conditions
column SPB-608, 30 m x 0.25 mm I.D., 0.25 µm (24103-U)
oven 100 C (2 min), 15 C/min to 300 C (5 min)
carrier gas helium, 0.9 ml/min, constant flow
sample chlorinated pesticide standard (cat. No. 46845-U), 50 ppb in methylene chloride
injection 2.0 µL, splitless (0.75 min)
liner 4 mm I.D. single taper
inj. temp. 250 C
detector ECD, 300 C
Description
Categories Analytical Chromatography, GC Applications, Pesticides
Featured Industry Environmental
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,1,1-Trichloro-2,2-bis(4-chlorophenyl)ethane (4,4′-DDT)
  • 1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane (4,4′-DDD)
  • 2,4,5,6-Tetrachloro-m-xylene
  • Aldrin
  • Decachlorobiphenyl
  • Dieldrin
  • Endosulfan I
  • Endosulfan II
  • Endosulfan sulfate
  • Endrin
  • Endrin aldehyde
  • Endrin ketone
  • Heptachlor
  • Heptachlor epoxide
  • Methoxychlor
  • α-1,2,3,4,5,6-Hexachlorocyclohexane
  • α-BHC
  • α-Chlordane
  • β-BHC
  • γ-BHC
  • γ-Chlordane
  • δ-BHC