US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column SLB-5ms, 15 m x 0.10 mm I.D., 0.10 µm (28466-U)
oven 100 °C, 25 °C/min to 325 °C
carrier gas hydrogen, 40 cm/sec constant
sample 50 ppb of a 22 component chlorinated pesticide standard in n-hexane
injection 2 µL, splitless (0.75 min)
liner 4 mm I.D., single taper
inj. temp. 225 °C
detector ECD, 300 °C
Description
Categories Analytical Chromatography, GC Applications, Pesticides
Featured Industry Environmental
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane (4,4′-DDD)
  • 2,4,5,6-Tetrachloro-m-xylene
  • 4,4′-DDE (1,1-Dichloro-2,2-bis(p-chlorophenyl)ethylene)
  • 4,4′-DDT
  • Aldrin
  • Decachlorobiphenyl
  • Dieldrin
  • Endosulfan sulfate
  • Endrin
  • Endrin aldehyde
  • Endrin ketone
  • Heptachlor
  • Heptachlor exo-epoxide
  • Lindane
  • Methoxychlor
  • α-1,2,3,4,5,6-Hexachlorocyclohexane
  • α-Chlordane
  • α-Endosulfan
  • β-BHC
  • β-Endosulfan
  • γ-Chlordane
  • δ-BHC