US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-35ms, 340 °C Final Temp.
US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-35ms, 340 °C Final Temp.
Conditions
column SLB-35ms, 30 m x 0.25 mm I.D., 0.25 µm (29804-U)
oven 100 °C (2 min), 15 °C/min to 340 °C (5 min)
carrier gas helium, 0.9 mL/min
sample 200 ppm organochlorine pesticide standard mix (CRM46845) containing 22 analytes, diluted to 50 ppb with hexane
injection 1µL, splitless (0.75 min)
liner 4 mm I.D., split/splitless type, wool packed single taper FocusLiner™ design
inj. temp. 250 °C
detector muECD, 340 °C
Description
Analysis Note The analysis of low levels of 20 organochlorine pesticides and 2 surrogate compounds by gas chromatography-electron capture detector (GC-ECD) is routinely performed by analysts in environmental laboratories. Methods require the analysis of each sample extract on two columns with differing selectivity for confirmation of identifications. The SLB-5ms/SLB-35ms pair is an effective column set because all 22 analytes are resolved between both columns using the same run conditions.
This application shows the separation achieved on SLB-35ms. Peaks are well integrated and baselines are exceptionally stable considering the high final oven temperature (340 ºC). Minimal baseline rise is detected, and the last analyte, decachlorobiphenyl, is eluted in less than 20 minutes.
Categories Analytical Chromatography, GC Applications, Pesticides
Featured Industry Environmental
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC