Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
US EPA Method 624: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap
US EPA Method 624: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using
Conditions
sample/matrix each analyte at 50 ppb in 5 mL water
purge trap VOCARB 3000 "K"
purge 40 ml/min at 25 C for 11 min
dry purge 2 min
desorption preheat 205 C
desorption process 40 ml/min at 210 C for 2 min
bake 260 C for 10 min
column SPB-624, 30 m x 0.25 mm I.D., 1.4 µm (24255)
oven 40 C (2 min), 7 C/min to 135 C, 30 C/min to 230 C (3 min)
carrier gas helium, 1.1 ml/min
injection 30:1 split
liner 0.75 mm I.D. SPME
inj. temp. 150 C
transfer line/valve temp. 110 C
MSD interface 200 C
scan range m/z = 35-400
Description
Categories Analytical Chromatography, SPME Applications, GC Applications, Environmental, Volatiles
Featured Industry Environmental
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,1,1-Trichloroethane
  • 1,1,2,2-Tetrachloroethane
  • 1,1,2-Trichloroethane
  • 1,1-Dichloroethane
  • 1,1-Dichloroethylene
  • 1,2-Dichlorobenzene
  • 1,2-Dichloroethane
  • 1,2-Dichloroethane-d4
  • 1,2-Dichloropropane
  • 1,3-Dichlorobenzene
  • 1,4-Dichlorobenzene
  • 1,4-Dichlorobenzene-d4
  • 2-Chloroethylvinylether
  • 4-Bromofluorobenzene
  • Benzene
  • Bromodichloromethane
  • Bromoform
  • Bromomethane
  • Carbon tetrachloride
  • Chlorobenzene
  • Chlorobenzene-d5
  • Chloroethane
  • Chloroform
  • Chloromethane
  • cis-1,3-Dichloropropene
  • Dibromochloromethane
  • Dibromofluoromethane
  • Dichloromethane
  • Ethylbenzene
  • Fluorobenzene
  • Tetrachloroethylene
  • Toluene
  • Toluene-d8
  • trans-1,2-Dichloroethylene
  • trans-1,3-Dichloropropene
  • Trichloroethylene
  • Trichlorofluoromethane
  • Vinyl chloride