US EPA Method 8015: GC Analysis of Nonhalogenated Volatiles on SLB®-5ms
US EPA Method 8015: GC Analysis of Nonhalogenated Volatiles on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 1.0 µm (28476-U)
oven 50 °C (3 min), 15 °C/min to 200 °C
carrier gas helium, 1.3 mL/min., constant
sample each analyte at 2000 µg/mL in methanol
injection 0.5 µL50:1 split
liner 2 mm I.D., FocusLiner™ inlet liner
inj. temp. 150 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Volatiles
Featured Industry Environmental
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 2-Butanone
  • 4-Methyl-2-pentanone
  • Ethanol
  • Ethyl ether
  • Methanol