US EPA Method 8015 (modified): GC Analysis of Diesel Range Organics (DRO) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
US EPA Method 8015 (modified): GC Analysis of Diesel Range Organics (DRO) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Conditions
column SLB-5ms, 20 m x 0.18 mm I.D., 0.18 µm (28564-U)
oven 50 °C (2 min), 20 °C/min to 325 °C (1 min)
carrier gas helium, 0.9 mL/min, constant
sample each analyte at 50 µg/mL in methylene chloride
injection 1 µL, splitless (splitter open at 0.5 min)
liner 2 mm I.D., FocusLiner™ inlet liner
inj. temp. 250 °C
detector FID, 350 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Environmental
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 5-α-Androstane
  • Decane
  • Docosane
  • Dodecane
  • Eicosane
  • Hexacosane
  • Hexadecane
  • Methylene chloride
  • Octacosane
  • Octadecane
  • o-Terphenyl
  • Tetracosane
  • Tetradecane