US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on SLB®-5ms, Direct Injection
US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on SLB®-5ms, Direct Injection
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 1.0 µm (28476-U)
oven 50 °C (3 min), 15 °C/min to 200 °C
carrier gas helium, 1.3 mL/min., constant
sample EPA GRO Mix, 9 analytes at various concentrations in methanol (47577-U)
injection 0.5 µL, 50:1 split
liner 2 mm I.D., FocusLiner™ inlet liner
inj. temp. 150 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Volatiles, Hydrocarbons
Featured Industry Environmental
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes