Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
US EPA Method 8041: GC Analysis of Phenols on Equity®-5
US EPA Method 8041: GC Analysis of Phenols on Equity®-5
Conditions
column Equity-5, 30 m x 0.25 mm I.D., 0.25 µm (25312)
oven 40 C, 8 C/min to 300C
carrier gas helium, 25 cm/sec @ 60 C
sample 10 µg/mL each component (47899)
injection 1.0 µl 100:1 split
liner split, cup design
inj. temp. 225 C
detector FID, 340 C
Description
Categories Analytical Chromatography, GC Applications, Phenols
Featured Industry Environmental
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 2,3,4,5-Tetrachlorophenol
  • 2,3,4,6-Tetrachlorophenol
  • 2,3,4-Trichlorophenol
  • 2,3,5,6-Tetrachlorophenol
  • 2,3,5-Trichlorophenol
  • 2,4,5-Trichlorophenol
  • 2,4,6-Tribromophenol
  • 2,4,6-Trichlorophenol
  • 2,4-Dichlorophenol
  • 2,4-Dimethylphenol
  • 2,4-Dinitrophenol
  • 2,6-Dichlorophenol
  • 2-Bromophenol
  • 2-Chlorophenol
  • 2-Methyl-4,6-dinitrophenol
  • 2-Methylphenol
  • 2-Nitrophenol
  • 3-Methylphenol
  • 4-Chloro-3-methylphenol
  • 4-Methylphenol
  • 4-Nitrophenol
  • Pentachlorophenol
  • Pentafluorophenol
  • Phenol