Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
US EPA Method 8081: GC Analysis of Organochlorine Pesticides on Equity®-5
US EPA Method 8081: GC Analysis of Organochlorine Pesticides on Equity®-5
Conditions
column Equity(R)-5, 30 m x 0.25 mm I.D., 0.25 µm (28089-U)
oven 100 C (2 min), 15 C/min to 160 C, 5 C/min to 300 C (10 min)
carrier gas helium, 30 cm/sec @ 100 C
sample 50 ppb of a 22-component chlorinated pesticides standard (46845-U)
injection 2 µL, splitless (0.5 min)
liner 4 mm I.D., double taper
inj. temp. 225 C
detector ECD, 310 C
Description
Categories Analytical Chromatography, GC Applications, Pesticides
Featured Industry Environmental
General description As per United States Environmental Protection Agency (EPA), method 8081 may be used in determining the concentrations of organochlorine pesticides in solid and liquid extracts. It can be achieved by using fused-silica, open tubular, capillary columns. This method allows choosing between single column and a dual column.
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 2,4,5,6-Tetrachloro-m-xylene
  • 4,4′-DDD
  • 4,4′-DDE (1,1-Dichloro-2,2-bis(p-chlorophenyl)ethylene)
  • 4,4′-DDT
  • Aldrin
  • Decachlorobiphenyl
  • Dieldrin
  • Endosulfan I
  • Endosulfan II
  • Endosulfan sulfate
  • Endrin
  • Endrin aldehyde
  • Endrin ketone
  • Heptachlor
  • Heptachlor epoxide
  • Methoxychlor
  • α-BHC
  • α-Chlordane
  • β-BHC
  • γ-BHC
  • γ-Chlordane
  • δ-BHC