US EPA Method 8082: GC Analysis of Aroclor 1016/1260 (PCBs) on SLB®-5ms
US EPA Method 8082: GC Analysis of Aroclor 1016/1260 (PCBs) on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 100 °C (2 min), 15 °C/min to 330 °C (3 min)
carrier gas helium, 25 cm/sec, constant
sample Aroclor standard mix 1 (46846-U) diluted to 500 ppb / 50 ppb (Aroclors / surrogates) in n-hexane
injection 2 µLsplitless (0.75 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
detector ECD, 330 °C
Description
Analysis Note Aroclors are commercial mixtures of PCB congeners. Due to their stability, they ave been used in many different industrial and commercial applications. Because of their toxicity and ability to bioaccuµLate, production of these materials ceased in 1977. This application demonstrates the separation of two common Aroclor mixtures, Aroclor 1016 and Aroclor 1260 on the SLB-5ms.
Categories Analytical Chromatography, GC Applications, Dioxins/PCBs/PBDEs
Featured Industry Environmental
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • Aroclor 1016
  • Aroclor 1260