US EPA Method 8082: GC Analysis of Aroclor 1016/1260 (PCBs) on Equity®-1701 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
US EPA Method 8082: GC Analysis of Aroclor 1016/1260 (PCBs) on Equity®-1701 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column Equity-1701, 15 m x 0.10 mm I.D., 0.10 µm (28343-U)
oven 90 °C, 35 °C/min to 280 °C (3 min)
carrier gas hydrogen, 50 cm/sec constant
sample Aroclor standard mix 1 (46846-U) diluted to 200 ppb / 20 ppb(Aroclors / surrogates) in n-hexane
injection 2 µL, splitless (0.75 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
detector ECD, 280 °C
Description
Categories Analytical Chromatography, GC Applications, Dioxins/PCBs/PBDEs
Featured Industry Environmental
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 2,4,5,6-Tetrachloro-m-xylene
  • Aroclor 1016
  • Aroclor 1260
  • Decachlorobiphenyl