US EPA Method 8082: GC Analysis of Aroclor 1248 (PCB) on SLB®-5ms and SPB®-608
US EPA Method 8082: GC Analysis of Aroclor 1248 (PCB) on SLB®-5ms and SPB®-608
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
column SPB-608, 30 m x 0.25 mm I.D., 0.25 µm (24103-U)
oven 100 °C (2 min), 15 °C/min to 300 °C (10 min)
carrier gas helium, 30 cm/sec, constant
sample Ar1248 (44807) and surrogates (48460) at 500 ppb and 50 ppb in n-hexane
injection 2 µL, splitless (0.75 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
detector mu-ECD, 330 °C
Description
Categories Analytical Chromatography, GC Applications, Dioxins/PCBs/PBDEs
Featured Industry Environmental
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC, SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Aroclor 1248
  • Decachlorobiphenyl
  • Tetrachloro-m-xylene