Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
US EPA Method 8082: GC Analysis of Aroclor 1254 (PCB) on SLB®-5ms and Equity®-1701 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
US EPA Method 8082: GC Analysis of Aroclor 1254 (PCB) on SLB®-5ms and Equity®-1701 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column Equity-1701, 15 m x 0.10 mm I.D., 0.10 µm (28343-U)
column SLB-5ms, 15 m x 0.10 mm I.D., 0.10 µm (28466-U)
oven 80 C (0.5 min), 30 C/min to 280 C (3 min)
carrier gas hydrogen, 40 cm/sec, constant
sample Ar1254 (44808) and surrogates (48460) at 500 ppb and 50 ppb in n-hexane
injection 1 µL, splitless (0.75 min)
liner 4 mm I.D., single taper
inj. temp. 250 C
detector mu-ECD, 300 C
Description
Categories Analytical Chromatography, GC Applications, Dioxins/PCBs/PBDEs
Featured Industry Environmental
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC, SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Aroclor 1254
  • Decachlorobiphenyl
  • Tetrachloro-m-xylene