US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608
US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608
Conditions
column SPB-608, 30 m x 0.25 mm I.D., 0.25 µm (24103-U)
oven 80 °C (2 min), 15 °C/min to 250 °C, 8 °C/min to 300 °C (35 min)
carrier gas helium, 1.2 mL/min. constant
sample each analyte at 10 µg/mL in methylene chloride
injection 0.5 µL, splitless (1 min)
inj. temp. 300 °C
MSD interface 300 °C
scan range m/z 40 - 450
Description
Categories Analytical Chromatography, GC Applications, PAHs, Hydrocarbons
Featured Industry Environmental
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1-Fluoronaphthalene
  • 2-Fluorobiphenyl
  • 3-Methylcholanthrene
  • 7H-Dibenzo[c,g]carbazole
  • Acenaphthene
  • Acenaphthylene
  • Anthracene
  • Benzo[a]anthracene
  • Benzo[a]pyrene
  • Benzo[b]fluoranthene
  • Benzo[g,h,i]perylene
  • Benzo[j]fluoranthene
  • Benzo[k]fluoranthene
  • Carbazole
  • Chrysene
  • Dibenzo[a,e]pyrene
  • Dibenzo[a,h]acridine
  • Dibenzo[a,h]anthracene
  • Dibenzo[a,h]pyrene
  • Dibenzo[a,i]pyrene
  • Dibenzo[a,j]acridine
  • Fluoranthene
  • Fluorene
  • Indeno[1,2,3-cd]pyrene
  • Naphthalene
  • Phenanthrene
  • Pyrene