US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms
US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 80 °C (2 min), 15 °C/min to 250 °C, 8 °C/min to 300 °C (5 min)
carrier gas helium, 1.2 mL/min constant
sample each analyte at 10 µg/mL in methylene chloride
injection 0.5 µLsplitless (1 min)
liner 2 mm I.D., FocusLiner™ with taper
inj. temp. 300 °C
MSD interface 300 °C
scan range m/z 40 - 450
Description
Categories Analytical Chromatography, GC Applications, PAHs, Hydrocarbons
Featured Industry Environmental
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1-Fluoronaphthalene
  • 2-Fluorobiphenyl
  • 3-Methylcholanthrene
  • 7H-Dibenzo[c,g]carbazole
  • Acenaphthene
  • Acenaphthylene
  • Anthracene
  • Benzo[a]anthracene
  • Benzo[a]pyrene
  • Benzo[b]fluoranthene
  • Benzo[g,h,i]perylene
  • Benzo[j]fluoranthene
  • Benzo[k]fluoranthene
  • Carbazole
  • Chrysene
  • Dibenzo[a,e]pyrene
  • Dibenzo[a,h]acridine
  • Dibenzo[a,h]anthracene
  • Dibenzo[a,h]pyrene
  • Dibenzo[a,i]pyrene
  • Dibenzo[a,j]acridine
  • Fluoranthene
  • Fluorene
  • Indeno[1,2,3-cd]pyrene
  • Naphthalene
  • Phenanthrene
  • Pyrene