US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SPB®-608
US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SPB®-608
Conditions
column SPB-608, 30 m x 0.25 mm I.D., 0.25 µm (24103-U)
oven 60 °C (1 min), 5 °C/min to 200 °C, 15 °C/min to 300 °C (5 min)
carrier gas helium, 1.5 mL/min
sample nitrogen-phosphorous pesticide standard, each analyte at 100 ppb in MTBE
injection 2.0 µL, splitless (0.75 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
detector NPD, 320 °C
Description
Categories Analytical Chromatography, GC Applications, Pesticides
Featured Industry Environmental
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1-Bromo-2-nitrobenzene
  • Aspon
  • Atrazine
  • Bolstar
  • Carbophenothion
  • Chlorfenvinphos
  • Chloropyrifos
  • Coumaphos
  • Crotoxyphos
  • DEF
  • Demeton-O
  • Diazinon
  • Dichlorofenthion
  • Dichlorvos
  • Dicrotophos
  • Dimethoate
  • Dioxathion
  • Disulfoton
  • EPN
  • Ethion
  • Ethoprop
  • Ethyl azinphos
  • Ethyl parathion
  • Famphur
  • Fenitrothion
  • Fensulfothion
  • Fenthion
  • Fonofos
  • Hexamethyl phosphoramide (HMPA)
  • Leptophos
  • Malathion
  • Methyl azinphos
  • Methyl chloropyrifos
  • Methyl parathion
  • Mevinphos isomers
  • Monocrotophos
  • Naled
  • Phorate
  • Phosmet
  • Phosphamidon
  • Ronnel
  • Simazine
  • Stirofos (Tetrachlorvinphos)
  • Sulfotep
  • Terbufos
  • Tetraethyl pyrophosphate (TEPP)
  • Thionazin (Zinophos)
  • Tokuthion
  • Tributyl phosphate
  • Trichlorfon
  • Trichloronate
  • Tri-o-cresyl phosphate (TOCP)
  • Triphenyl phosphate