Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
US EPA Method 8260: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis
US EPA Method 8260: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using
Conditions
sample/matrix each analyte at 50 ppb in 5 mL water
purge trap VOCARB 3000 K (24940-U)
purge 40 ml/min at 25 C for 11 min
dry purge 2 min
desorption preheat 205 C
desorption temp. 210 ºC for 1 min
desorption flow 150 ml/min
bake 260 C for 10 min
column SPB-624, 20 m x 0.18 mm I.D., 1.0 µm (28662-U)
oven 40 C (1 min), 11 C/min to 125 C, 35 C/min to 230 C (2 min)
carrier gas helium, 1.5 ml/min
injection 100:1 split
liner 0.75 mm I.D. SPME
inj. temp. 150 C
transfer line/valve temp. 110 C
MSD interface 220 C
scan range m/z = 35-400
Description
Analysis Note US EPA Method 8260 describes the analysis of volatile organic compounds in solid wastes and ground waters. This application illustrates the analysis of many compounds commonly analyzed by this method using purge and trap coupled to GC-MS. A short, narrow I.D. SPB-624 column was used to shorten the analysis time to under 11 minutes. A high desorb flow through the trap was necessary to prevent band broadening of the gaseous VOCs. This was accomplished by increasing the split ratio, thus increasing the total flow through the trap during desorb.
Categories Analytical Chromatography, SPME Applications, GC Applications, Volatiles, Environmental
Featured Industry Environmental
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,1,1,2-Tetrachloroethane
  • 1,1,1-Trichloroethane
  • 1,1,2,2-Tetrachloroethane
  • 1,1,2-Trichloroethane
  • 1,1-Dichloroethane
  • 1,1-Dichloroethene
  • 1,1-Dichloropropene
  • 1,2,3-Trichlorobenzene
  • 1,2,3-Trichloropropane
  • 1,2,4-Trichlorobenzene
  • 1,2,4-Trimethylbenzene
  • 1,2-Dibromo-3-chloropropane
  • 1,2-Dibromomethane
  • 1,2-Dichlorobenzene
  • 1,2-Dichloroethane
  • 1,2-Dichloroethane-d4
  • 1,2-Dichloropropane
  • 1,3,5-Trimethylbenzene
  • 1,3-Dichlorobenzene
  • 1,3-Dichloropropane
  • 1,4-Dichlorobenzene
  • 1,4-Dichlorobenzene-d4
  • 2,2-Dichloropropane
  • 2-Butanone
  • 2-Hexanone
  • 4-Bromofluorobenzene
  • 4-Methyl-2-pentanone
  • Acetone
  • Benzene
  • Bromobenzene
  • Bromochloromethane
  • Bromodichloromethane
  • Bromoform
  • Bromomethane
  • Butylbenzene
  • Carbon tetrachloride
  • Chlorobenzene
  • Chlorobenzene-d5
  • Chloroethane
  • Chloroform
  • Chloromethane
  • cis-1,2-Dichloroethene
  • cis-1,3-Dichloropropene
  • cis-1,4-Dichloro-2-butene
  • Dibromochloromethane
  • Dibromofluoromethane
  • Dibromomethane
  • Dichlorofluoromethane
  • Ethylbenzene
  • Fluorobenzene
  • Hexachlorobutadiene
  • Iodomethane
  • Isopropylbenzene (cumene)
  • Methylene chloride
  • m-Xylene
  • Naphthalene
  • n-Propylbenzene
  • o-Xylene
  • p-Chlorotoluene
  • Pentachloroethane
  • p-Isopropyltoluene
  • p-Xylene
  • sec-Butylbenzene
  • Styrene
  • tert-Butylbenzene
  • Tetrachloroethene
  • Toluene
  • Toluene-d8
  • trans-1,2-Dichloroethene
  • trans-1,3-Dichloropropene
  • trans-1,4-Dichloro-2-butene
  • Trichloroethene
  • Trichlorofluoromethane
  • Vinyl chloride
  • α-Chlorotoluene